The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281495.1 | NP_001268424.1 | 15 - 16 | 1802 | 67 |
NM_030877.4 | NP_110517.2 | 14 - 15 | 1701 | 67 | |
XM_011528917.2 | XP_011527219.1 | 12 - 13 | 1312 | 67 | |
GenBank mRNA | AF239607.1 | - | 14 - 15 | 1627 | 67 |
AF367471.1 | - | 11 - 12 | 1218 | 67 | |
AF370431.1 | - | 14 - 15 | 1607 | 67 | |
AK024761.1 | - | 5 - 6 | 555 | 67 | |
AK074663.1 | - | 14 - 15 | 1614 | 67 | |
AK091216.1 | - | 14 - 15 | 2715 | 67 | |
AK130301.1 | - | 14 - 15 | 1614 | 67 | |
AK222592.1 | - | 5 - 6 | 555 | 67 | |
AK293420.1 | - | 15 - 16 | 1695 | 67 | |
BC022802.1 | - | 6 - 7 | 718 | 67 | |
BC036739.1 | - | 7 - 8 | 864 | 67 | |
BC121005.1 | - | 14 - 15 | 1622 | 67 | |
BC121006.1 | - | 14 - 15 | 1622 | 67 |