The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018054.2 | NP_001018064.1 | 11 - 12 | 2009 | 90 |
NM_004631.4 | NP_004622.2 | 11 - 12 | 2009 | 90 | |
NM_017522.4 | NP_059992.3 | 10 - 11 | 1622 | 90 | |
NM_033300.3 | NP_150643.2 | 9 - 10 | 1499 | 90 | |
XM_005271173.3 | XP_005271230.1 | 12 - 13 | 1958 | 90 | |
XM_005271174.3 | XP_005271231.1 | 11 - 12 | 1566 | 90 | |
XM_005271175.3 | XP_005271232.1 | 10 - 11 | 1527 | 90 | |
XM_006710881.3 | XP_006710944.1 | 12 - 13 | 1958 | 90 | |
XM_006710882.3 | XP_006710945.1 | 12 - 13 | 1958 | 90 | |
XM_011542094.2 | XP_011540396.1 | 11 - 12 | 1832 | 90 | |
XM_011542095.2 | XP_011540397.1 | 10 - 11 | 1437 | 90 | |
XM_011542096.2 | XP_011540398.1 | 12 - 13 | 1958 | 90 | |
XM_017002265.1 | XP_016857754.1 | 10 - 11 | 1793 | 90 | |
XM_017002266.1 | XP_016857755.1 | 12 - 13 | 1958 | 90 | |
XM_017002267.1 | XP_016857756.1 | 10 - 11 | 1403 | 90 | |
XM_017002268.1 | XP_016857757.1 | 11 - 12 | 1442 | 90 | |
GenBank mRNA | AK096482.1 | - | 9 - 10 | 1277 | 90 |
AK122887.1 | - | 3 - 4 | 2469 | 90 | |
AL119002.1 | - | 4 - 5 | 661 | 90 | |
BC051836.1 | - | 11 - 12 | 1914 | 90 | |
CD300289.1 | - | 1 - 2 | 122 | 90 | |
D50678.1 | - | 11 - 12 | 1874 | 90 | |
Z75190.1 | - | 10 - 11 | 1382 | 90 |