Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAKMIP1
Entrez Gene ID
152789
Gene Name
janus kinase and microtubule interacting protein 1
Gene Aliases
Gababrbp, JAMIP1, MARLIN1
Chromosome Location
Chr.4: 6024925 - 6200591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099433.1 NP_001092903.1 8 - 9 1786 78
NM_001306133.1 NP_001293062.1 8 - 9 1576 78
NM_001306134.1 NP_001293063.1 7 - 8 1081 78
NM_144720.3 NP_653321.1 8 - 9 1786 78
XM_011513400.2 XP_011511702.1 7 - 8 1255 78
XM_017007788.1 XP_016863277.1 8 - 9 1750 78
XM_017007789.1 XP_016863278.1 8 - 9 1574 78
XM_017007790.1 XP_016863279.1 8 - 9 1726 78
XM_017007791.1 XP_016863280.1 8 - 9 1750 78
XM_017007792.1 XP_016863281.1 8 - 9 1750 78
XM_017007793.1 XP_016863282.1 8 - 9 1750 78
XM_017007794.1 XP_016863283.1 8 - 9 1750 78
XM_017007795.1 XP_016863284.1 8 - 9 1750 78
XM_017007796.1 XP_016863285.1 8 - 9 1750 78
XM_017007797.1 XP_016863286.1 7 - 8 1255 78
XM_017007798.1 XP_016863287.1 7 - 8 1893 78
XM_017007799.1 XP_016863288.1 7 - 8 1255 78
GenBank mRNA AK056126.1 - 8 - 9 1750 78
AK098536.1 - 6 - 7 860 78
AK124718.1 - 8 - 9 1574 78
AK294356.1 - 9 - 10 1403 78
AK295339.1 - 7 - 8 1081 78
AK307983.1 - 7 - 8 1001 78
AM412307.1 - 8 - 9 1750 78
AM412309.1 - 8 - 9 1750 78
AY382340.1 - 7 - 8 1300 78
BC047075.1 - 8 - 9 1506 78

View Full Product Details