Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRSF4
Entrez Gene ID
6429
Gene Name
serine and arginine rich splicing factor 4
Gene Aliases
SFRS4, SRP75
Chromosome Location
Chr.1: 29147738 - 29182125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK310528.1 - 1 - 2 242 75

View Full Product Details