The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024669.2 | NP_078945.2 | 3 - 4 | 333 | 84 |
XM_017009852.1 | XP_016865341.1 | 2 - 3 | 218 | 84 | |
XM_017009853.1 | XP_016865342.1 | 2 - 3 | 217 | 84 | |
GenBank mRNA | AK123389.1 | - | 3 - 4 | 333 | 84 |
DA292581.1 | - | 3 - 4 | 284 | 84 |