The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261411.1 | NP_001248340.1 | 13 - 14 | 2281 | 84 |
NM_004592.3 | NP_004583.2 | 13 - 14 | 2281 | 84 | |
XM_011538653.1 | XP_011536955.1 | 12 - 13 | 2047 | 84 | |
XM_011538654.2 | XP_011536956.1 | 13 - 14 | 2233 | 84 | |
XM_011538655.2 | XP_011536957.1 | 13 - 14 | 2278 | 84 | |
XM_017019798.1 | XP_016875287.1 | 12 - 13 | 2047 | 84 | |
XM_017019799.1 | XP_016875288.1 | 13 - 14 | 2720 | 84 | |
GenBank mRNA | AB208921.1 | - | 11 - 12 | 5472 | 84 |
BC008707.2 | - | 13 - 14 | 2278 | 84 | |
BC015953.2 | - | 13 - 14 | 2278 | 84 | |
BC136678.1 | - | 13 - 14 | 2231 | 84 | |
BC144364.1 | - | 13 - 14 | 2231 | 84 | |
U08377.1 | - | 13 - 14 | 2258 | 84 |