Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SFSWAP
Entrez Gene ID
6433
Gene Name
splicing factor SWAP homolog
Gene Aliases
SFRS8, SWAP
Chromosome Location
Chr.12: 131711087 - 131799738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261411.1 NP_001248340.1 17 - 18 3012 145
NM_004592.3 NP_004583.2 16 - 17 2856 145
XM_011538653.1 XP_011536955.1 16 - 17 2778 145
XM_011538654.2 XP_011536956.1 17 - 18 2964 145
XM_017019798.1 XP_016875287.1 15 - 16 2622 145
XM_017019799.1 XP_016875288.1 17 - 18 3451 145
GenBank mRNA BC136678.1 - 16 - 17 2806 145
BC144364.1 - 17 - 18 2962 145
U08377.1 - 16 - 17 2833 145

View Full Product Details