The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308392.1 | NP_001295321.1 | 12 - 13 | 2451 | 91 |
GenBank mRNA | D21255.1 | - | 12 - 13 | 2372 | 91 |