The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022756.5 | NP_073593.2 | 5 - 6 | 603 | 125 |
NR_073090.1 | 5 - 6 | 603 | 183 | ||
NR_073092.1 | 6 - 7 | 692 | 125 | ||
GenBank mRNA | AK308118.1 | - | 5 - 6 | 537 | 125 |
AK309663.1 | - | 6 - 7 | 653 | 125 | |
BX538212.1 | - | 5 - 6 | 539 | 125 | |
BX640719.1 | - | 5 - 6 | 548 | 125 |