Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
Chromosome Location
Chr.8: 37736579 - 37757801 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007175.6 NP_009106.1 7 - 8 618 75
XM_005273392.2 XP_005273449.1 7 - 8 778 75
XM_006716280.2 XP_006716343.1 6 - 7 523 75
GenBank mRNA AK291394.1 - 7 - 8 571 75
AL442077.1 - - 605 75
AY358108.1 - 7 - 8 562 75
BC005950.1 - 7 - 8 541 75

View Full Product Details