Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERLIN2
Entrez Gene ID
11160
Gene Name
ER lipid raft associated 2
Gene Aliases
C8orf2, Erlin-2, NET32, SPFH2, SPG18
Chromosome Location
Chr.8: 37736579 - 37757801 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007175.6 NP_009106.1 9 - 10 765 56
XM_005273392.2 XP_005273449.1 9 - 10 925 56
XM_006716280.2 XP_006716343.1 8 - 9 670 56
GenBank mRNA AK291394.1 - 9 - 10 718 56
AL442077.1 - - 752 56
AY358108.1 - 9 - 10 709 56
BC005950.1 - 9 - 10 688 56

View Full Product Details