Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPBG
Entrez Gene ID
7162
Gene Name
trophoblast glycoprotein
Gene Aliases
5T4, 5T4AG, M6P1, WAIF1
Chromosome Location
Chr.6: 82363206 - 82370828 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166392.1 NP_001159864.1 2 - 2 103
NM_006670.4 NP_006661.1 3 - 3 103
XM_011536097.2 XP_011534399.1 2 - 2 103
GenBank mRNA AI342491.1 - 1 - 1 276 103
AI857997.1 - 1 - 1 273 103

View Full Product Details