The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040214.2 | NP_001035304.1 | 5 - 6 | 887 | 87 |
NM_001300737.1 | NP_001287666.1 | 6 - 7 | 951 | 87 | |
NM_001300738.1 | NP_001287667.1 | 6 - 7 | 901 | 87 | |
NM_001300740.1 | NP_001287669.1 | 5 - 6 | 525 | 87 | |
NM_153355.4 | NP_699186.2 | 4 - 5 | 686 | 87 | |
XM_011535501.2 | XP_011533803.1 | 5 - 6 | 775 | 87 | |
XM_011535502.2 | XP_011533804.1 | 5 - 6 | 542 | 87 | |
XM_011535503.2 | XP_011533805.1 | 4 - 5 | 812 | 87 | |
XM_017010318.1 | XP_016865807.1 | 4 - 5 | 810 | 87 | |
XM_017010319.1 | XP_016865808.1 | 4 - 5 | 574 | 87 | |
GenBank mRNA | AK055326.1 | - | 5 - 6 | 520 | 87 |
AK308927.1 | - | 6 - 7 | 539 | 87 | |
AK309385.1 | - | 6 - 7 | 500 | 87 | |
AK309407.1 | - | 2 - 3 | 262 | 87 | |
AK309802.1 | - | 6 - 7 | 539 | 87 | |
AK310763.1 | - | 4 - 5 | 361 | 87 | |
AK315886.1 | - | 5 - 6 | 526 | 87 | |
BC035062.2 | - | 6 - 7 | 653 | 87 | |
DB174077.1 | - | 3 - 4 | 361 | 87 |