Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAX1BP1
Entrez Gene ID
8887
Gene Name
Tax1 binding protein 1
Gene Aliases
CALCOCO3, T6BP, TXBP151
Chromosome Location
Chr.7: 27739373 - 27829767 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006024.6 NP_006015.4 13 - 14 1933 145
XM_017012754.1 XP_016868243.1 13 - 14 1933 145
GenBank mRNA AK000098.1 - 13 - 14 145
AK314292.1 - 13 - 14 1903 145
AL136586.1 - 13 - 14 1839 145
BC050358.2 - 13 - 14 1818 145
CR533556.1 - 12 - 13 1751 145

View Full Product Details