Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNF76
Entrez Gene ID
7629
Gene Name
zinc finger protein 76
Gene Aliases
D6S229E, ZNF523, Zfp523
Chromosome Location
Chr.6: 35259473 - 35295987 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292032.1 NP_001278961.1 8 - 9 977 72
NM_003427.4 NP_003418.2 8 - 9 977 72
XM_011514846.2 XP_011513148.1 8 - 9 987 72
XM_011514847.2 XP_011513149.1 8 - 9 1125 72
XM_011514849.2 XP_011513151.1 8 - 9 961 72
XM_011514850.2 XP_011513152.1 8 - 9 1089 72
XM_011514854.2 XP_011513156.1 8 - 9 974 72
XM_011514855.2 XP_011513157.1 8 - 9 1102 72
XM_011514856.2 XP_011513158.1 5 - 6 1114 72
XM_017011248.1 XP_016866737.1 8 - 9 1125 72
XM_017011249.1 XP_016866738.1 8 - 9 1089 72
XM_017011251.1 XP_016866740.1 8 - 9 1125 72
XM_017011252.1 XP_016866741.1 8 - 9 1125 72
XM_017011253.1 XP_016866742.1 8 - 9 1089 72
XM_017011254.1 XP_016866743.1 8 - 9 961 72
XM_017011255.1 XP_016866744.1 8 - 9 1102 72
XM_017011256.1 XP_016866745.1 8 - 9 1104 72
XM_017011257.1 XP_016866746.1 8 - 9 1102 72
GenBank mRNA AB208784.1 - 7 - 8 1828 72
AK294605.1 - 8 - 9 961 72
AK295276.1 - 8 - 9 1079 72
AK302903.1 - 8 - 9 1079 72
BC000133.2 - 8 - 9 957 72
BC002549.2 - 8 - 9 958 72
BT007316.1 - 7 - 8 753 72
M91592.1 - 8 - 9 916 72

View Full Product Details