Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAMP3
Entrez Gene ID
10067
Gene Name
secretory carrier membrane protein 3
Gene Aliases
C1orf3
Chromosome Location
Chr.1: 155255979 - 155262703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005698.3 NP_005689.2 6 - 7 911 104
NM_052837.2 NP_443069.1 5 - 6 833 104
XM_006711105.3 XP_006711168.1 6 - 7 1187 104
XM_006711106.3 XP_006711169.1 4 - 5 1028 104
GenBank mRNA AF005039.1 - 6 - 7 773 104
BC000161.2 - 6 - 7 785 104
BC005135.2 - 6 - 7 758 104
BC010505.1 - 5 - 6 702 104

View Full Product Details