Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BIN3
Entrez Gene ID
55909
Gene Name
bridging integrator 3
Gene Aliases
-
Chromosome Location
Chr.8: 22620418 - 22669148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018688.4 NP_061158.1 7 - 8 572 111
XM_005273573.3 XP_005273630.1 6 - 7 478 111
XM_011544586.2 XP_011542888.1 8 - 9 657 111
GenBank mRNA AF271732.1 - 7 - 8 535 111
AK001289.1 - 5 - 6 434 111
AK023980.1 - 7 - 8 519 111
AK124486.1 - 1 - 2 857 111
AK222470.1 - 7 - 8 544 111
BC001223.1 - 6 - 7 456 111
BC009824.2 - 7 - 8 506 111

View Full Product Details