The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018688.4 | NP_061158.1 | 5 - 6 | 393 | 61 |
XM_005273573.3 | XP_005273630.1 | 4 - 5 | 299 | 61 | |
XM_011544586.2 | XP_011542888.1 | 6 - 7 | 478 | 61 | |
XM_011544587.1 | XP_011542889.1 | 5 - 6 | 393 | 61 | |
GenBank mRNA | AF271732.1 | - | 5 - 6 | 356 | 61 |
AK001289.1 | - | 3 - 4 | 255 | 61 | |
AK023980.1 | - | 5 - 6 | 340 | 61 | |
AK090446.1 | - | 2 - 3 | 902 | 61 | |
AK222470.1 | - | 5 - 6 | 365 | 61 | |
BC001223.1 | - | 4 - 5 | 277 | 61 | |
BC009824.2 | - | 5 - 6 | 327 | 61 |