The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207035.1 | NP_001193964.1 | 1 - 2 | 264 | 89 |
NM_001207036.1 | NP_001193965.1 | 1 - 2 | 264 | 89 | |
NM_001207037.1 | NP_001193966.1 | 1 - 2 | 264 | 89 | |
NM_001207038.1 | NP_001193967.1 | 1 - 2 | 255 | 89 | |
NM_001207039.1 | NP_001193968.1 | 1 - 2 | 264 | 89 | |
NM_016135.3 | NP_057219.1 | 1 - 2 | 264 | 89 | |
GenBank mRNA | AF116508.1 | - | 1 - 2 | 80 | 89 |
AF116509.1 | - | 1 - 2 | 80 | 89 | |
AF147782.1 | - | 1 - 2 | 181 | 89 | |
AF218235.1 | - | 1 - 2 | 80 | 89 | |
AF218366.1 | - | 1 - 2 | 80 | 89 | |
AJ276205.1 | - | 1 - 2 | 196 | 89 | |
AK026144.1 | - | 1 - 2 | 248 | 89 | |
AK122796.1 | - | 1 - 2 | 864 | 89 | |
AK223424.1 | - | 1 - 2 | 183 | 89 | |
AK303829.1 | - | 1 - 2 | 256 | 89 | |
BC035853.1 | - | 1 - 2 | 154 | 89 |