The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207035.1 | NP_001193964.1 | 6 - 7 | 1064 | 73 |
NM_001207036.1 | NP_001193965.1 | 5 - 6 | 899 | 73 | |
NM_001207037.1 | NP_001193966.1 | 6 - 7 | 1105 | 73 | |
NM_001207038.1 | NP_001193967.1 | 5 - 6 | 824 | 73 | |
NM_001207039.1 | NP_001193968.1 | 5 - 6 | 899 | 73 | |
NM_001207040.1 | NP_001193969.1 | 5 - 6 | 928 | 73 | |
NM_001207041.1 | NP_001193970.1 | 4 - 5 | 763 | 73 | |
NM_016135.3 | NP_057219.1 | 6 - 7 | 1064 | 73 | |
XM_011514659.1 | XP_011512961.1 | 6 - 7 | 945 | 73 | |
GenBank mRNA | AF116508.1 | - | 6 - 7 | 921 | 73 |
AF116509.1 | - | 6 - 7 | 880 | 73 | |
AF116510.1 | - | 5 - 6 | 744 | 73 | |
AF147782.1 | - | 5 - 6 | 750 | 73 | |
AF175387.3 | - | 6 - 7 | 73 | ||
AF218235.1 | - | 5 - 6 | 715 | 73 | |
AF218365.1 | - | 6 - 7 | 830 | 73 | |
AF218366.1 | - | 5 - 6 | 715 | 73 | |
AJ276205.1 | - | 6 - 7 | 996 | 73 | |
AK026144.1 | - | 6 - 7 | 1048 | 73 | |
AK122796.1 | - | 6 - 7 | 1664 | 73 | |
AK223424.1 | - | 6 - 7 | 983 | 73 | |
AK301011.1 | - | 4 - 5 | 680 | 73 | |
AK303829.1 | - | 5 - 6 | 891 | 73 | |
BC035853.1 | - | 6 - 7 | 954 | 73 |