Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGAP2
Entrez Gene ID
116986
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTG1, GGAP2, PIKE
Chromosome Location
Chr.12: 57723765 - 57742201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122772.2 NP_001116244.1 5 - 6 1547 142
NM_014770.3 NP_055585.1 5 - 6 629 142
XM_005268625.3 XP_005268682.1 5 - 6 1673 142
XM_005268626.2 XP_005268683.1 5 - 6 669 142
XM_017018772.1 XP_016874261.1 5 - 6 1674 142
GenBank mRNA AF384128.1 - 5 - 6 625 142
AF413077.1 - 5 - 6 539 142
AK122827.1 - 5 - 6 633 142
AK292672.1 - 5 - 6 601 142
AY128689.1 - 5 - 6 1547 142
BC028020.1 - 5 - 6 596 142
D79989.1 - 5 - 6 627 142

View Full Product Details