Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGAP2
Entrez Gene ID
116986
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTG1, GGAP2, PIKE
Chromosome Location
Chr.12: 57723765 - 57742201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122772.2 NP_001116244.1 7 - 8 1795 75
NM_014770.3 NP_055585.1 7 - 8 877 75
XM_005268625.3 XP_005268682.1 7 - 8 1921 75
XM_005268626.2 XP_005268683.1 7 - 8 917 75
XM_017018772.1 XP_016874261.1 7 - 8 1922 75
GenBank mRNA AF384128.1 - 7 - 8 873 75
AF413077.1 - 7 - 8 787 75
AK122827.1 - 7 - 8 881 75
AK292672.1 - 7 - 8 849 75
AY128689.1 - 7 - 8 1795 75
BC028020.1 - 7 - 8 844 75
D79989.1 - 7 - 8 875 75

View Full Product Details