The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172695.1 | NP_001166166.1 | 3 - 4 | 421 | 93 |
NM_001172696.1 | NP_001166167.1 | 3 - 4 | 421 | 93 | |
NM_001172697.1 | NP_001166168.1 | 3 - 4 | 421 | 93 | |
NM_005726.5 | NP_005717.3 | 3 - 4 | 421 | 93 | |
GenBank mRNA | AF110399.1 | - | 3 - 4 | 367 | 93 |
AK295323.1 | - | 5 - 6 | 653 | 93 | |
AK304621.1 | - | 3 - 4 | 405 | 93 | |
AK308981.1 | - | 3 - 4 | 384 | 93 | |
AK313453.1 | - | 3 - 4 | 405 | 93 | |
BC022862.1 | - | 3 - 4 | 394 | 93 | |
BC093068.1 | - | 3 - 4 | 376 | 93 | |
L37936.1 | - | 2 - 3 | 272 | 93 |