The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291965.1 | NP_001278894.1 | 4 - 5 | 496 | 80 |
NM_006646.5 | NP_006637.2 | 4 - 5 | 496 | 80 | |
XM_011534889.1 | XP_011533191.1 | 4 - 5 | 507 | 80 | |
XM_011534890.1 | XP_011533192.1 | 3 - 4 | 437 | 80 | |
GenBank mRNA | AB020707.2 | - | 3 - 4 | 385 | 80 |
AB026543.1 | - | 2 - 3 | 271 | 80 | |
AF134305.1 | - | 1 - 2 | 130 | 80 | |
AF454702.1 | - | 4 - 5 | 449 | 80 | |
AK289845.1 | - | 4 - 5 | 496 | 80 | |
BC050283.1 | - | 4 - 5 | 496 | 80 |