Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFS1
Entrez Gene ID
7466
Gene Name
wolframin ER transmembrane glycoprotein
Gene Aliases
CTRCT41, WFRS, WFS, WFSL
Chromosome Location
Chr.4: 6260368 - 6303265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145853.1 NP_001139325.1 4 - 5 626 75
NM_006005.3 NP_005996.2 4 - 5 630 75
XM_017008586.1 XP_016864075.1 5 - 6 609 75
GenBank mRNA AF084481.1 - 4 - 5 630 75
AK312897.1 - 4 - 5 559 75
BC030130.2 - 4 - 5 560 75
KU178395.1 - 3 - 4 460 75
Y18064.1 - 4 - 5 618 75

View Full Product Details