Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RMND5B
Entrez Gene ID
64777
Gene Name
required for meiotic nuclear division 5 homolog B
Gene Aliases
GID2, GID2B
Chromosome Location
Chr.5: 178130532 - 178148570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288794.1 NP_001275723.1 10 - 11 1351 116
NM_001288795.1 NP_001275724.1 9 - 10 1200 116
NM_022762.4 NP_073599.2 9 - 10 1211 116
XM_005265969.3 XP_005266026.1 8 - 9 1031 116
XM_005265971.4 XP_005266028.1 9 - 10 1778 116
XM_005265972.3 XP_005266029.1 8 - 9 1158 116
XM_017009727.1 XP_016865216.1 8 - 9 1822 116
XM_017009728.1 XP_016865217.1 8 - 9 1489 116
GenBank mRNA AK025971.1 - 6 - 7 756 116
AK093481.1 - 9 - 10 1152 116
AK093622.1 - 1 - 2 1668 116
AK094065.1 - 6 - 7 1358 116
AK122692.1 - 4 - 5 2074 116
AK125337.1 - 10 - 11 1298 116
AL137381.1 - 3 - 4 324 116
AL713670.1 - 8 - 9 1074 116
AY359092.1 - 6 - 7 2069 116
BC009911.2 - 9 - 10 1145 116
DQ494789.1 - 9 - 10 1145 116

View Full Product Details