Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWC25
Entrez Gene ID
54883
Gene Name
CWC25 spliceosome associated protein homolog
Gene Aliases
CCDC49
Chromosome Location
Chr.17: 38800434 - 38825350 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017748.4 NP_060218.1 1 - 2 180 60
NR_073428.1 1 - 2 180 60
GenBank mRNA AK000298.1 - 1 - 2 130 60
AK225373.1 - 1 - 2 151 60
AK296115.1 - 1 - 2 145 60
BC003085.1 - 1 - 2 33 60
BC008833.2 - 1 - 2 166 60
CR749362.1 - 1 - 2 151 60
HY021361.1 - 1 - 2 181 60

View Full Product Details