The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145855.1 | NP_001139327.1 | 1 - 2 | 256 | 66 |
GenBank mRNA | AK299996.1 | - | 1 - 2 | 172 | 66 |
AK304752.1 | - | 1 - 2 | 158 | 66 | |
DC391267.1 | - | 1 - 2 | 257 | 66 |