The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015425.4 | NP_056240.2 | 33 - 34 | 5442 | 77 |
GenBank mRNA | AK025568.1 | - | 10 - 11 | 1621 | 77 |
AK302458.1 | - | 33 - 34 | 5167 | 77 | |
AK304131.1 | - | 20 - 21 | 3419 | 77 | |
AL117467.1 | - | 2 - 3 | 189 | 77 | |
BC017115.1 | - | 10 - 11 | 1627 | 77 | |
BC117173.1 | - | 33 - 34 | 5095 | 77 | |
BC126303.1 | - | 33 - 34 | 5095 | 77 | |
BC143345.1 | - | 33 - 34 | 5095 | 77 | |
U33460.1 | - | 33 - 34 | 5124 | 77 |