The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015044.4 | NP_055859.1 | 12 - 13 | 1239 | 64 |
XM_017023074.1 | XP_016878563.1 | 13 - 14 | 4928 | 64 | |
XM_017023075.1 | XP_016878564.1 | 12 - 13 | 1242 | 64 | |
GenBank mRNA | AB029003.1 | - | 9 - 10 | 869 | 64 |
AF165531.1 | - | 12 - 13 | 1177 | 64 | |
AF190863.1 | - | 12 - 13 | 1183 | 64 | |
AF233522.1 | - | 12 - 13 | 1182 | 64 | |
AF323754.1 | - | 13 - 14 | 1319 | 64 | |
AK024619.1 | - | 12 - 13 | 1238 | 64 | |
AK308425.1 | - | 12 - 13 | 1226 | 64 | |
BC000284.1 | - | 12 - 13 | 1221 | 64 |