The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015044.4 | NP_055859.1 | 15 - 16 | 1698 | 94 |
XM_017023074.1 | XP_016878563.1 | 16 - 17 | 5387 | 94 | |
XM_017023075.1 | XP_016878564.1 | 15 - 16 | 1701 | 94 | |
GenBank mRNA | AB029003.1 | - | 12 - 13 | 1328 | 94 |
AF165531.1 | - | 15 - 16 | 1636 | 94 | |
AF190863.1 | - | 15 - 16 | 1642 | 94 | |
AF233522.1 | - | 15 - 16 | 1641 | 94 | |
AK024619.1 | - | 15 - 16 | 1697 | 94 | |
BC000284.1 | - | 15 - 16 | 1680 | 94 |