The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015044.4 | NP_055859.1 | 2 - 3 | 262 | 79 |
XM_017023074.1 | XP_016878563.1 | 3 - 4 | 3951 | 79 | |
GenBank mRNA | AF165531.1 | - | 2 - 3 | 200 | 79 |
AF190863.1 | - | 2 - 3 | 206 | 79 | |
AF233522.1 | - | 2 - 3 | 205 | 79 | |
AF323754.1 | - | 2 - 3 | 204 | 79 | |
AK024619.1 | - | 2 - 3 | 262 | 79 | |
AK308425.1 | - | 2 - 3 | 249 | 79 | |
BC000284.1 | - | 2 - 3 | 244 | 79 | |
DA439738.1 | - | 2 - 3 | 262 | 79 |