Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGA2
Entrez Gene ID
23062
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
VEAR
Chromosome Location
Chr.16: 23463542 - 23510494 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015044.4 NP_055859.1 7 - 8 737 69
XM_017023074.1 XP_016878563.1 8 - 9 4426 69
XM_017023075.1 XP_016878564.1 7 - 8 740 69
GenBank mRNA AB029003.1 - 4 - 5 367 69
AF165531.1 - 7 - 8 675 69
AF190863.1 - 7 - 8 681 69
AF233522.1 - 7 - 8 680 69
AF323754.1 - 8 - 9 819 69
AK024619.1 - 7 - 8 736 69
BC000284.1 - 7 - 8 719 69

View Full Product Details