The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020718.3 | NP_065769.3 | 9 - 10 | 1625 | 106 |
XM_017023496.1 | XP_016878985.1 | 9 - 10 | 1656 | 106 | |
XM_017023497.1 | XP_016878986.1 | 9 - 10 | 3151 | 106 | |
XM_017023498.1 | XP_016878987.1 | 8 - 9 | 938 | 106 | |
XM_017023499.1 | XP_016878988.1 | 6 - 7 | 742 | 106 | |
XM_017023500.1 | XP_016878989.1 | 9 - 10 | 1656 | 106 | |
GenBank mRNA | AJ586135.2 | - | 9 - 10 | 1625 | 106 |
AK126752.1 | - | 9 - 10 | 106 |