The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020718.3 | NP_065769.3 | 10 - 11 | 1688 | 148 |
XM_017023496.1 | XP_016878985.1 | 10 - 11 | 1719 | 148 | |
XM_017023497.1 | XP_016878986.1 | 10 - 11 | 3214 | 148 | |
XM_017023498.1 | XP_016878987.1 | 9 - 10 | 1001 | 148 | |
XM_017023499.1 | XP_016878988.1 | 7 - 8 | 805 | 148 | |
XM_017023500.1 | XP_016878989.1 | 10 - 11 | 1719 | 148 | |
GenBank mRNA | BM842261.1 | - | 4 - 5 | 385 | 148 |