The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284351.2 | NP_001271280.1 | 15 - 16 | 2338 | 70 |
NM_004174.3 | NP_004165.2 | 15 - 16 | 2365 | 70 | |
GenBank mRNA | BC101669.1 | - | 15 - 16 | 2261 | 70 |
BC101671.1 | - | 15 - 16 | 2261 | 70 | |
BC143328.1 | - | 15 - 16 | 2234 | 70 | |
U28043.1 | - | 15 - 16 | 2263 | 70 |