Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBXN2A
Entrez Gene ID
165324
Gene Name
UBX domain protein 2A
Gene Aliases
UBXD4
Chromosome Location
Chr.2: 23927201 - 24001000 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181713.3 NP_859064.2 2 - 3 201 143
XM_005264166.4 XP_005264223.1 2 - 3 473 143
XM_005264167.4 XP_005264224.1 2 - 3 176 143
XM_005264168.4 XP_005264225.1 3 - 4 610 143
XM_005264169.4 XP_005264226.1 2 - 3 199 143
XM_011532633.2 XP_011530935.1 3 - 4 596 143
XM_017003517.1 XP_016859006.1 2 - 3 261 143
GenBank mRNA AK291192.1 - 3 - 4 518 143
AL832079.1 - 2 - 3 202 143
BC092484.1 - 2 - 3 292 143
BC093681.1 - 2 - 3 101 143
BC111997.1 - 2 - 3 113 143
BC143298.1 - 2 - 3 101 143

View Full Product Details