The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001809.3 | NP_001800.1 | 3 - 4 | 475 | 67 |
GenBank mRNA | BC002703.2 | - | 3 - 4 | 346 | 67 |
BT007246.1 | - | 3 - 4 | 292 | 67 | |
U14518.1 | - | 3 - 4 | 435 | 67 |