The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170795.1 | NP_001164266.1 | 1 - 2 | 116 | 116 |
NM_080592.3 | NP_542159.3 | 1 - 2 | 436 | 116 | |
GenBank mRNA | AB009017.1 | - | 1 - 2 | 129 | 116 |
AF077037.1 | - | 1 - 2 | 118 | 116 | |
AY358968.1 | - | 1 - 2 | 109 | 116 | |
BC002846.1 | - | 1 - 2 | 104 | 116 | |
BC011006.1 | - | 1 - 2 | 110 | 116 | |
BC035850.1 | - | 1 - 2 | 102 | 116 | |
BQ718371.1 | - | 1 - 2 | 151 | 116 |