Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRGPRX2
Entrez Gene ID
117194
Gene Name
MAS related GPR family member X2
Gene Aliases
MGRG3, MRGX2
Chromosome Location
Chr.11: 19054455 - 19060681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054030.3 NP_473371.1 1 - 2 61 71
GenBank mRNA BC063450.1 - 1 - 2 61 71

View Full Product Details