The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321378.1 | NP_001308307.1 | 13 - 14 | 2511 | 151 |
NM_001321380.1 | NP_001308309.1 | 11 - 12 | 1312 | 151 | |
NM_002645.3 | NP_002636.2 | 12 - 13 | 2442 | 151 | |
XM_017017924.1 | XP_016873413.1 | 13 - 14 | 2458 | 151 | |
GenBank mRNA | AK294416.1 | - | 11 - 12 | 1299 | 151 |
BC113658.1 | - | 11 - 12 | 2294 | 151 | |
BX648778.1 | - | 2 - 3 | 378 | 151 | |
Y13367.1 | - | 11 - 12 | 2228 | 151 |