The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321378.1 | NP_001308307.1 | 14 - 15 | 2672 | 126 |
NM_001321380.1 | NP_001308309.1 | 12 - 13 | 1473 | 126 | |
NM_002645.3 | NP_002636.2 | 13 - 14 | 2603 | 126 | |
XM_017017924.1 | XP_016873413.1 | 14 - 15 | 2619 | 126 | |
GenBank mRNA | AK294416.1 | - | 12 - 13 | 1460 | 126 |
BC113658.1 | - | 12 - 13 | 2455 | 126 | |
BX648778.1 | - | 3 - 4 | 539 | 126 |