The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321378.1 | NP_001308307.1 | 23 - 24 | 3827 | 95 |
NM_001321380.1 | NP_001308309.1 | 21 - 22 | 2628 | 95 | |
NM_002645.3 | NP_002636.2 | 22 - 23 | 3758 | 95 | |
XM_011520186.2 | XP_011518488.1 | 21 - 22 | 3585 | 95 | |
XM_017017924.1 | XP_016873413.1 | 23 - 24 | 3774 | 95 | |
GenBank mRNA | AK294416.1 | - | 21 - 22 | 2615 | 95 |
AK309610.1 | - | 5 - 6 | 518 | 95 | |
BC113658.1 | - | 21 - 22 | 3610 | 95 | |
BX648778.1 | - | 12 - 13 | 1694 | 95 | |
Y13367.1 | - | 21 - 22 | 3544 | 95 |