Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM2
Entrez Gene ID
2515
Gene Name
ADAM metallopeptidase domain 2
Gene Aliases
CRYN1, CRYN2, CT15, FTNB, PH-30b, PH30, PH30-beta
Chromosome Location
Chr.8: 39743735 - 39838289 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278114.1 NP_001265043.1 7 - 8 679 113
NM_001464.4 NP_001455.3 7 - 8 679 113
XM_005273468.1 XP_005273525.1 7 - 8 617 113
GenBank mRNA AJ133005.1 - 7 - 8 579 113
AK301734.1 - 7 - 8 617 113
U52370.1 - 7 - 8 582 113
X99374.1 - 7 - 8 575 113

View Full Product Details