The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007203.4 | NP_009134.1 | 6 - 7 | 690 | 86 |
NM_147150.2 | NP_671492.1 | 6 - 7 | 690 | 86 | |
GenBank mRNA | AB023137.1 | - | 6 - 7 | 690 | 86 |
AJ303079.1 | - | 6 - 7 | 678 | 86 | |
BC140818.1 | - | 6 - 7 | 531 | 86 |