The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282588.1 | NP_001269517.1 | 3 - 4 | 685 | 93 |
NM_001282589.1 | NP_001269518.1 | 4 - 5 | 789 | 93 | |
NM_001282590.1 | NP_001269519.1 | 5 - 6 | 861 | 93 | |
NM_001282591.1 | NP_001269520.1 | 4 - 5 | 741 | 93 | |
NM_001282592.1 | NP_001269521.1 | 5 - 6 | 1078 | 93 | |
NM_001282593.1 | NP_001269522.1 | 3 - 4 | 637 | 93 | |
NM_001282594.1 | NP_001269523.1 | 5 - 6 | 900 | 93 | |
NM_001282595.1 | NP_001269524.1 | 5 - 6 | 643 | 93 | |
NM_001282596.1 | NP_001269525.1 | 4 - 5 | 757 | 93 | |
NM_021181.4 | NP_067004.3 | 6 - 7 | 1182 | 93 | |
XM_011509828.1 | XP_011508130.1 | 6 - 7 | 1031 | 93 | |
XM_011509829.1 | XP_011508131.1 | 5 - 6 | 927 | 93 | |
GenBank mRNA | AB027233.1 | - | 6 - 7 | 952 | 93 |
AF291815.1 | - | 6 - 7 | 948 | 93 | |
AF390894.1 | - | 6 - 7 | 957 | 93 | |
AJ271869.1 | - | 5 - 6 | 91 | ||
AJ276429.2 | - | 6 - 7 | 970 | 93 | |
AK290706.1 | - | 5 - 6 | 862 | 93 | |
AK292097.1 | - | 5 - 6 | 645 | 93 | |
AK292148.1 | - | 6 - 7 | 966 | 93 | |
AK298499.1 | - | 4 - 5 | 596 | 93 | |
AK298548.1 | - | 3 - 4 | 493 | 93 | |
AK301137.1 | - | 3 - 4 | 428 | 93 | |
AK301432.1 | - | 5 - 6 | 686 | 93 | |
AK301438.1 | - | 4 - 5 | 529 | 93 | |
AL713801.1 | - | 1 - 2 | 1807 | 93 | |
AL834424.1 | - | 5 - 6 | 650 | 93 | |
AY358512.1 | - | 6 - 7 | 974 | 93 | |
BC027867.1 | - | 5 - 6 | 844 | 93 | |
DA644251.1 | - | 4 - 5 | 478 | 93 |