The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166285.1 | NP_001159757.1 | 3 - 4 | 397 | 65 |
NM_006429.3 | NP_006420.1 | 2 - 3 | 301 | 65 | |
NR_029402.1 | 2 - 3 | 301 | 65 | ||
XM_011532478.2 | XP_011530780.1 | 3 - 4 | 331 | 65 | |
XM_011532479.1 | XP_011530781.1 | 2 - 3 | 234 | 65 | |
GenBank mRNA | AF026292.1 | - | 2 - 3 | 226 | 65 |
AK098549.1 | - | 2 - 3 | 219 | 65 | |
AK222478.1 | - | 2 - 3 | 209 | 65 | |
AK291961.1 | - | 2 - 3 | 247 | 65 | |
AK293175.1 | - | 2 - 3 | 219 | 65 | |
AK297846.1 | - | 3 - 4 | 325 | 65 | |
BC019296.1 | - | 2 - 3 | 214 | 65 | |
BC088351.1 | - | 2 - 3 | 256 | 65 | |
BM918058.1 | - | - | 277 | 65 | |
CR456719.1 | - | 1 - 2 | 158 | 65 | |
CR536511.1 | - | 1 - 2 | 158 | 65 | |
DC362157.1 | - | 2 - 3 | 301 | 65 | |
DC395264.1 | - | 2 - 3 | 299 | 65 | |
U83843.1 | - | 1 - 2 | 122 | 65 |