The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001232.3 | NP_001223.2 | 3 - 4 | 674 | 67 |
GenBank mRNA | AK129891.1 | - | 3 - 4 | 649 | 67 |
AK225959.1 | - | 3 - 4 | 536 | 67 | |
AK293595.1 | - | 3 - 4 | 683 | 67 | |
AK313041.1 | - | 3 - 4 | 620 | 67 | |
BC022288.1 | - | 3 - 4 | 583 | 67 | |
D55655.1 | - | 3 - 4 | 519 | 67 |