Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL4A6
Entrez Gene ID
1288
Gene Name
collagen type IV alpha 6 chain
Gene Aliases
CXDELq22.3, DELXq22.3, DFNX6
Chromosome Location
Chr.X: 108155607 - 108439497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287758.1 NP_001274687.1 34 - 35 3561 75
NM_001287759.1 NP_001274688.1 33 - 34 3510 75
NM_001287760.1 NP_001274689.1 33 - 34 3510 75
NM_001847.3 NP_001838.2 33 - 34 3405 75
NM_033641.3 NP_378667.1 33 - 34 3510 75
XM_006724617.3 XP_006724680.1 34 - 35 3450 75
XM_011530852.2 XP_011529154.1 34 - 35 3450 75
XM_011530853.2 XP_011529155.1 32 - 33 4166 75
XM_011530854.2 XP_011529156.1 34 - 35 3451 75
GenBank mRNA BC140855.1 - 33 - 34 3406 75
BC144669.1 - 33 - 34 3406 75
CA489342.1 - 3 - 4 306 75
D21337.1 - 33 - 34 3510 75
U04845.1 - 33 - 34 3388 75

View Full Product Details