The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015360.4 | NP_056175.3 | 18 - 19 | 2320 | 147 |
GenBank mRNA | AK291649.1 | - | 18 - 19 | 2067 | 147 |
AK304788.1 | - | 16 - 17 | 1862 | 147 | |
BC014669.2 | - | 9 - 10 | 1048 | 147 | |
BC028604.2 | - | 18 - 19 | 2078 | 147 | |
BC031779.1 | - | 6 - 7 | 718 | 147 | |
BC065258.1 | - | 18 - 19 | 2059 | 147 | |
BC104996.1 | - | 18 - 19 | 2099 | 147 | |
BC113509.1 | - | 18 - 19 | 2099 | 147 | |
BX640789.1 | - | 18 - 19 | 2280 | 147 | |
D29641.2 | - | 18 - 19 | 2066 | 147 |